KEY087886935 Key Organics / BIONET

N-(3-chloro-4-methylphenyl)methanesulfonamide

  • MDL number MFCD01212924
  • Cas Number : 71270-61-6
  • Molecular Weight : 219.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94845

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-0049_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart