KEY087886940 Key Organics / BIONET

6-amino-1,3-diethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD00086294
  • Cas Number : 41740-15-2
  • Molecular Weight : 183.211

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94849

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-0055_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart