KEY087886947 Key Organics / BIONET

methyl 7-methoxy-2,2-diphenyl-2H-1,3-benzodioxole-5-carboxylate

  • MDL number MFCD00183318
  • Cas Number : 102706-14-9
  • Molecular Weight : 362.381

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94856

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-0062_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart