KEY087886953 Key Organics / BIONET

N-{4-[(2-cyanophenyl)methoxy]phenyl}acetamide

  • MDL number MFCD06672957
  • Cas Number : 852386-93-7
  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144 - 146
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70065

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2Z-0804_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart