KEY087886954 Key Organics / BIONET

6-amino-5-benzoyl-1-methyl-1,2-dihydropyrimidine-2-thione

  • MDL number MFCD02947011
  • Cas Number : 210235-77-1
  • Molecular Weight : 245.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237 - 240
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70067

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2Z-0812_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart