KEY087886956 Key Organics / BIONET

5-amino-1-benzyl-1H-1,2,3-triazole-4-carboxamide

  • MDL number MFCD00514988
  • Molecular Weight : 217.232

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97625

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/H-34005_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart