KEY087886958 Key Organics / BIONET

3-benzyl-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-ol

  • MDL number MFCD02942199
  • Cas Number : 21324-31-2
  • Molecular Weight : 227.227

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >250
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97627

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/H-38001_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart