KEY090495445 Key Organics / BIONET

methyl 1-(benzenesulfonyl)-4-bromo-1H-pyrrole-2-carboxylate

  • MDL number MFCD08064739
  • Cas Number : 882747-46-8
  • Molecular Weight : 344.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137 - 139
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73803

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3Y-0821_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart