KEY090496735 Key Organics / BIONET

1-(4-bromo-1-methyl-1H-pyrrol-2-yl)-2,2,2-trichloroethan-1-one

  • MDL number MFCD08276938
  • Cas Number : 184643-69-4
  • Molecular Weight : 305.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102 - 104
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3696

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10Z-0815_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart