KEY090496737 Key Organics / BIONET

3-(3-bromophenyl)-5-methyl-1H-pyrazole

  • MDL number MFCD08276940
  • Cas Number : 324054-75-3
  • Molecular Weight : 237.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101 - 104
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3698

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10Z-0827_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart