KEY090496741 Key Organics / BIONET

2-amino-1-(4-bromophenyl)ethan-1-ol

  • MDL number MFCD08276931
  • Cas Number : 41147-82-4
  • Molecular Weight : 216.078

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111 - 113
Purity 95
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7383

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11Z-0710_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart