KEY090496745 Key Organics / BIONET

tert-butyl 4-(3-acetyl-4-nitrophenyl)piperazine-1-carboxylate

  • MDL number MFCD08276930
  • Cas Number : 886361-04-2
  • Molecular Weight : 349.387

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167 - 169
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7399

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11Z-0848_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart