KEY090496751 Key Organics / BIONET

methyl 7-amino-3-cyano-2-(cyanomethyl)pyrazolo[1,5-a]pyrimidine-6-carboxylate

  • MDL number MFCD08276962
  • Cas Number : 886361-35-9
  • Molecular Weight : 256.225

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 279 - 281DEC
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63118

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12Z-0709_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart