KEY090496752 Key Organics / BIONET

1-[2-amino-5-(4-hydroxypiperidin-1-yl)phenyl]ethan-1-one

  • MDL number MFCD08276932
  • Cas Number : 359841-43-3
  • Molecular Weight : 234.299

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184 - 186
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63119

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12Z-0800_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart