KEY090496754 Key Organics / BIONET

1-[2-amino-5-(4-methylpiperidin-1-yl)phenyl]ethan-1-one

  • MDL number MFCD08276955
  • Cas Number : 886361-39-3
  • Molecular Weight : 232.327

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79 - 81
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63128

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12Z-0826_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart