KEY090496756 Key Organics / BIONET

2-hydrazinyl-5-nitro-1H-1,3-benzodiazole

  • MDL number MFCD08276961
  • Cas Number : 91467-48-0
  • Molecular Weight : 193.166

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 235 - 236
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63134

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12Z-0922_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart