KEY090496760 Key Organics / BIONET

4-(tert-butylsulfanyl)benzaldehyde

  • MDL number MFCD08443944
  • Cas Number : 88357-16-8
  • Molecular Weight : 194.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77322

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4X-0731_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart