KEY090496761 Key Organics / BIONET

(2E)-2-[(E)-2-bromobenzoyl]-3-(dimethylamino)prop-2-enenitrile

  • MDL number MFCD08276956
  • Cas Number : 886361-83-7
  • Molecular Weight : 279.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 88 - 90
Iupac Name (2E)-2-(2-bromobenzoyl)-3-(dimethylamino)-2-propenenitrile
Inchi Code 1S/C12H11BrN2O/c1-15(2)8-9(7-14)12(16)10-5-3-4-6-11(10)13/h3-6,8H,1-2H3/b9-8+
Inchi Key YACLVWZVUUPOTD-CMDGGOBGSA-N

Description

Melting Point 88 - 90
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77527

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4Z-0827_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart