KEY090496762 Key Organics / BIONET

6-propyl-4-(trifluoromethyl)pyridin-2-ol

  • MDL number MFCD08443946
  • Cas Number : 937601-39-3
  • Molecular Weight : 205.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 75 - 7
Iupac Name 6-propyl-4-(trifluoromethyl)-2-pyridinol
Inchi Code 1S/C9H10F3NO/c1-2-3-7-4-6(9(10,11)12)5-8(14)13-7/h4-5H,2-3H2,1H3,(H,13,14)
Inchi Key KIPUZHQWADBGHG-UHFFFAOYSA-N

Description

Melting Point 75 - 7
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80960

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5W-0931_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart