KEY090496765 Key Organics / BIONET

4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]but-3-yn-2-ol

  • MDL number MFCD08443949
  • Cas Number : 937601-45-1
  • Molecular Weight : 249.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 155 @ 0.5 mBar
Iupac Name 4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-3-butyn-2-ol
Inchi Code 1S/C10H7ClF3NO/c1-6(16)2-3-9-8(11)4-7(5-15-9)10(12,13)14/h4-6,16H,1H3
Inchi Key KJCIABAWPAIOIT-UHFFFAOYSA-N

Description

Melting Point 155 @ 0.5 mBar
Purity 95%
Physical Form Low Melting Solid,Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84372

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6X-0835_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart