KEY090496782 Key Organics / BIONET

tert-butyl 4-(2-fluoro-4-nitrophenyl)piperazine-1-carboxylate

  • MDL number MFCD08443959
  • Cas Number : 154590-34-8
  • Molecular Weight : 325.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95428

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CA-0603_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart