KEY090496813 Key Organics / BIONET

ethyl 2-chloro-1,3-benzothiazole-6-carboxylate

  • MDL number MFCD08443995
  • Cas Number : 78485-37-7
  • Molecular Weight : 241.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90 - 92
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95519

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CA-0925_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart