KEY090496829 Key Organics / BIONET

O-(2-aminoethyl)hydroxylamine dihydrochloride

  • MDL number MFCD04117898
  • Cas Number : 37866-45-8
  • Molecular Weight : 149.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205 - 207
Purity 95
Storage temp. Store under N2 at -18
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71784

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P411,P422
hcodes H302,H312,H315,H319,H332,H335,H412
MSDS https://www.keyorganics.net/media/catalog/product/sds/3L-902_SDS.pdf
pictograms GHS07,GHS09
Availability details for
Quantity
  • Availability
  • Add to Cart