KEY090496839 Key Organics / BIONET

O-[(2-fluorophenyl)methyl]hydroxylamine hydrochloride

  • MDL number MFCD01114584
  • Cas Number : 215599-91-0
  • Molecular Weight : 177.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180 - 185
Purity 95
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74370

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4F-951_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart