KEY090496840 Key Organics / BIONET

O-[(3-fluorophenyl)methyl]hydroxylamine hydrochloride

  • MDL number MFCD01114585
  • Cas Number : 51572-90-8
  • Molecular Weight : 177.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 218 - 225
Iupac Name 1-[(aminooxy)methyl]-3-fluorobenzene hydrochloride
Inchi Code 1S/C7H8FNO.ClH/c8-7-3-1-2-6(4-7)5-10-9;/h1-4H,5,9H2;1H
Inchi Key OSOKLAPGVHYYPY-UHFFFAOYSA-N

Description

Melting Point 218 - 225
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74371

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4F-952_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart