KEY090496847 Key Organics / BIONET

1,2,3-thiadiazole-4-carbohydrazide

  • MDL number MFCD00708766
  • Cas Number : 4100-18-9
  • Molecular Weight : 144.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207 - 209
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6369

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11P-305S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart