KEY090496856 Key Organics / BIONET

5-(diphenylmethyl)-1H-pyrazole

  • MDL number MFCD00104842
  • Cas Number : 143547-74-4
  • Molecular Weight : 234.302

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89 - 90
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7080

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11T-0869_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart