KEY090496860 Key Organics / BIONET

tert-butyl 3-formyl-5-methoxy-1H-indole-1-carboxylate

  • MDL number MFCD05864693
  • Cas Number : 324756-80-1
  • Molecular Weight : 275.304

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138 - 140
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7377

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11Z-0701_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart