KEY090496864 Key Organics / BIONET

5-(thiophen-3-yl)-1H-pyrazole

  • MDL number MFCD00121014
  • Cas Number : 19933-25-6
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 112 - 114
Iupac Name 5-(3-thienyl)-1H-pyrazole
Inchi Code 1S/C7H6N2S/c1-3-8-9-7(1)6-2-4-10-5-6/h1-5H,(H,8,9)
Inchi Key UOAGMESLUAMTIP-UHFFFAOYSA-N

Description

Melting Point 112 - 114
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84461

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/6Y-0804_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart