KEY090496880 Key Organics / BIONET

2-(aminooxy)-1-(morpholin-4-yl)ethan-1-one hydrochloride

  • MDL number MFCD01114501
  • Cas Number : 312521-29-2
  • Molecular Weight : 196.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202 - 204
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92095

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9G-920_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart