KEY090496885 Key Organics / BIONET

N-(3,4-dichlorophenyl)acetamide

  • MDL number MFCD00018148
  • Cas Number : 2150-93-8
  • Molecular Weight : 204.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 121 - 124
Iupac Name N-(3,4-dichlorophenyl)acetamide
Inchi Code 1S/C8H7Cl2NO/c1-5(12)11-6-2-3-7(9)8(10)4-6/h2-4H,1H3,(H,11,12)
Inchi Key SCYGGCAQZFJGRF-UHFFFAOYSA-N

Description

Melting Point 121 - 124
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92466

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/9J-545S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart