KEY090496892 Key Organics / BIONET

2-tert-butyl-7-methyl-1H-indole

  • MDL number MFCD05224832
  • Cas Number : 69622-42-0
  • Molecular Weight : 187.286

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94347

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9Z-0810_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart