KEY090496897 Key Organics / BIONET

4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbohydrazide

  • MDL number MFCD00112085
  • Cas Number : 263387-25-3
  • Molecular Weight : 301.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170 - 173
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3678

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10Z-0701_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart