KEY090496915 Key Organics / BIONET

3-(4-methoxyphenyl)-1H-pyrazol-5-amine

  • MDL number MFCD00462192
  • Cas Number : 19541-95-8
  • Molecular Weight : 189.218

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143 - 145
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94409

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AA-0860_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart