KEY090496919 Key Organics / BIONET

3-(3,4-dimethoxyphenyl)-3-oxopropanenitrile

  • MDL number MFCD02260783
  • Cas Number : 4640-69-1
  • Molecular Weight : 205.213

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139 - 141
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94415

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AA-0868_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart