KEY090496924 Key Organics / BIONET

2-[3-(trifluoromethyl)phenyl]-1,3-thiazole-4-carbohydrazide

  • MDL number MFCD01936330
  • Cas Number : 400083-64-9
  • Molecular Weight : 287.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161 - 163
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3691

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10Z-0720_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart