KEY090496925 Key Organics / BIONET

3-(3,5-dimethoxyphenyl)-3-oxopropanenitrile

  • MDL number MFCD00158877
  • Cas Number : 70988-04-4
  • Molecular Weight : 205.213

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121 - 123
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94866

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BA-0800_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart