KEY090496936 Key Organics / BIONET

O-[(4-methylphenyl)methyl]hydroxylamine hydrochloride

  • MDL number MFCD01114570
  • Cas Number : 38936-62-8
  • Molecular Weight : 173.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220 - 230
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7751

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12E-920_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart