KEY090496948 Key Organics / BIONET

3-(2-chlorophenyl)-1H-pyrazole

  • MDL number MFCD00111882
  • Cas Number : 59843-55-9
  • Molecular Weight : 178.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76 - 77
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9885

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12T-0845_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart