KEY090496967 Key Organics / BIONET

2-(aminooxy)-1-{4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl}ethan-1-one hydrochloride

  • MDL number MFCD01114567
  • Cas Number : 321391-87-1
  • Molecular Weight : 375.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172 - 177
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4401

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11F-925_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart