KEY090496974 Key Organics / BIONET

5-tert-butyl-2-methoxybenzaldehyde

  • MDL number MFCD06246086
  • Cas Number : 85943-26-6
  • Molecular Weight : 192.258

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point low melt solid
Purity 95
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95491

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P422
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CA-0854_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart