KEY090496988 Key Organics / BIONET

O-[(2,5-dichlorophenyl)methyl]hydroxylamine hydrochloride

  • MDL number MFCD01114576
  • Cas Number : 317821-71-9
  • Molecular Weight : 228.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188 - 195
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63897

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1G-927_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart