KEY090496989 Key Organics / BIONET

O-[(2-nitrophenyl)methyl]hydroxylamine hydrochloride

  • MDL number MFCD01114577
  • Cas Number : 51572-92-0
  • Molecular Weight : 204.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164 - 170
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63898

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1G-928_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart