KEY090496992 Key Organics / BIONET

O-[(2-methoxyphenyl)methyl]hydroxylamine hydrochloride

  • MDL number MFCD01114579
  • Cas Number : 317821-72-0
  • Molecular Weight : 189.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 112 - 116
Iupac Name 1-[(aminooxy)methyl]-2-methoxybenzene hydrochloride
Inchi Code 1S/C8H11NO2.ClH/c1-10-8-5-3-2-4-7(8)6-11-9;/h2-5H,6,9H2,1H3;1H
Inchi Key VTZSQCSODFGNCD-UHFFFAOYSA-N

Description

Melting Point 112 - 116
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63907

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/1G-939_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart