KEY090496999 Key Organics / BIONET

5-methyl-4-(thiophen-2-yl)-1,3-thiazol-2-amine

  • MDL number MFCD02734128
  • Cas Number : 206555-52-4
  • Molecular Weight : 196.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109 - 111
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95967

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DA-0703_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart