KEY090497002 Key Organics / BIONET

7-amino-2H,5H,6H,7H-indeno[5,6-d][1,3]dioxol-5-one hydrochloride

  • MDL number MFCD04035629
  • Cas Number : 148502-20-9
  • Molecular Weight : 227.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 270 - 273DEC
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65827

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-0603_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart