KEY090497015 Key Organics / BIONET

(2E)-3-[4-(trifluoromethoxy)phenyl]prop-2-enoic acid

  • MDL number MFCD00066338
  • Cas Number : 783-13-1
  • Molecular Weight : 232.158

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185 - 186
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/98454

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-038C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart