KEY090497023 Key Organics / BIONET

methyl 3,5-dichloro-4-methoxybenzoate

  • MDL number MFCD00017189
  • Cas Number : 24295-27-0
  • Molecular Weight : 235.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74 - 77
Purity 95
Storage temp. Store under N2
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/99590

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-163C_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart