KEY092592846 Key Organics / BIONET

1-[(2-chloro-1,3-thiazol-5-yl)methyl]-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD03102582
  • Cas Number : 692287-27-7
  • Molecular Weight : 226.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58 - 60
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83958

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6T-0065_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart