KEY092592867 Key Organics / BIONET

methyl 4-[(4-methylphenyl)methoxy]benzoate

  • MDL number MFCD01568711
  • Cas Number : 62290-48-6
  • Molecular Weight : 256.301

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97 - 99
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93070

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9N-051_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart